Product Details

SNP ID
rs7651075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14652143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGAACCTGGTGTCCCGTGGGTT[A/G]GGAAACTCCGTGCCTCACCTCTGCC
Phenotype
MIM: 616735
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC174 PubMed Links

Gene Details

Gene
CCDC174
Gene Name
coiled-coil domain containing 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016474.4 Intron NP_057558.3
XM_017006554.1 Intron XP_016862043.1
XM_017006555.1 Intron XP_016862044.1

View Full Product Details