Product Details

SNP ID
rs4494157
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:27370346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATAGAGGAGGAAACTGAGGCTCA[G/T]TGGAGGGGTAGGACTTGCCCAAGGT
Phenotype
MIM: 604973 MIM: 604468
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD164L2 PubMed Links

Gene Details

Gene
CD164L2
Gene Name
CD164 molecule like 2
There are no transcripts associated with this gene.

Gene
FCN3
Gene Name
ficolin 3
There are no transcripts associated with this gene.

Gene
MAP3K6
Gene Name
mitogen-activated protein kinase kinase kinase 6
There are no transcripts associated with this gene.

View Full Product Details