Product Details

SNP ID
rs1572788
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:153931407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGAGATAGGTGCTTATGCCATTT[C/T]ACAGATGACCAAATTGAGGCTCAGA
Phenotype
MIM: 614998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND4B PubMed Links

Gene Details

Gene
DENND4B
Gene Name
DENN domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014856.2 Intron NP_055671.2
XM_005245678.4 Intron XP_005245735.1
XM_005245679.2 Intron XP_005245736.1
XM_011510219.1 Intron XP_011508521.1
XM_011510220.2 Intron XP_011508522.1
XM_011510221.2 Intron XP_011508523.1
XM_011510222.1 Intron XP_011508524.1
XM_017002993.1 Intron XP_016858482.1
Gene
GATAD2B
Gene Name
GATA zinc finger domain containing 2B
There are no transcripts associated with this gene.

View Full Product Details