Product Details

SNP ID
rs1956449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:41609710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAGGAGTGTTCGTTTGTCTCCC[C/G]CCTTTGGGAGCTGCTTGCTGGCTAC
Phenotype
MIM: 612811
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRFN5 PubMed Links

Gene Details

Gene
LRFN5
Gene Name
leucine rich repeat and fibronectin type III domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152447.3 Intron NP_689660.2
XM_005267369.3 Intron XP_005267426.1
XM_017021048.1 Intron XP_016876537.1
XM_017021049.1 Intron XP_016876538.1
XM_017021050.1 Intron XP_016876539.1

View Full Product Details