Product Details

SNP ID
rs2056086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15681200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAAATAAGCCTGAGAGTACACC[A/G]TTTCATTAAAGAGAGGAGAAAAGGC
Phenotype
MIM: 300631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CA5BP1 PubMed Links

Gene Details

Gene
CA5BP1
Gene Name
carbonic anhydrase 5B pseudogene 1
There are no transcripts associated with this gene.

Gene
TMEM27
Gene Name
transmembrane protein 27
There are no transcripts associated with this gene.

View Full Product Details