Product Details

SNP ID
rs15842
Assay Type
Functionally tested
NCBI dbSNP Submissions
76
Location
Chr.1:1013541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCATCTTTGCCAGTACAGGAGCT[C/T]GTGCCGTGGCCCACAGCCCACAGCC
Phenotype
MIM: 103320 MIM: 147571
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGRN PubMed Links

Gene Details

Gene
AGRN
Gene Name
agrin
There are no transcripts associated with this gene.

Gene
ISG15
Gene Name
ISG15 ubiquitin-like modifier
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005101.3 75 UTR 5 NP_005092.1

View Full Product Details