Product Details

SNP ID
rs28491407
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:1013673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTCCCAGGGGTGCAGGGAGAG[C/G]GGAGCTGCTCAGAGCTTGGCCAGGT
Phenotype
MIM: 103320 MIM: 147571
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AGRN PubMed Links

Gene Details

Gene
AGRN
Gene Name
agrin
There are no transcripts associated with this gene.

Gene
ISG15
Gene Name
ISG15 ubiquitin-like modifier
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005101.3 Intron NP_005092.1

View Full Product Details