Product Details

SNP ID
rs9892425
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:59164577 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAAGGAAGATGCTAGGAAATTTC[A/G]GATTCTAGGGCTCCCAATCAGTGGA
Phenotype
MIM: 615920 MIM: 616674
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRR11 PubMed Links

Gene Details

Gene
PRR11
Gene Name
proline rich 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018304.3 Intron NP_060774.2
Gene
SKA2
Gene Name
spindle and kinetochore associated complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details