Product Details

SNP ID
rs1863601
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:43203014 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTAAATGTAGCATTTTCTGGTAATC[C/T]TGCAGCACAGTCATCTGCTCTGAAG
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
There are no transcripts associated with this gene.

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 Intron NP_000110.2
NM_001114134.1 Intron NP_001107606.1
XM_005254225.1 Intron XP_005254282.1
XM_011521349.2 Intron XP_011519651.1
XM_011521350.2 Intron XP_011519652.1
XM_011521351.2 Intron XP_011519653.1
XM_011521352.2 Intron XP_011519654.1
XM_011521353.2 Intron XP_011519655.1
XM_011521354.2 Intron XP_011519656.1

View Full Product Details