Product Details
- SNP ID
-
rs2032778
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:15596573 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGTATGGGAAAGGTAACAAGATG[A/G]GCTGTAGTTGGAGTTGACAGAGGAA
- Phenotype
-
MIM: 601257
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX1
PubMed Links
Gene Details
- Gene
- DDX1
- Gene Name
- DEAD/H-box helicase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004939.2 |
|
Intron |
|
|
NP_004930.1 |
View Full Product Details