Product Details
- SNP ID
-
rs10919302
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
34
- Location
-
Chr.1:162382619 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGAGGTCACAAAGATAGTACTTG[A/G]CAGAGCTGGATTTGAACCCAGCTAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf111
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77061615] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1orf111
- Gene Name
- chromosome 1 open reading frame 111
There are no transcripts associated with this gene.
- Gene
- C1orf226
- Gene Name
- chromosome 1 open reading frame 226
View Full Product Details