Product Details

SNP ID
rs34540702
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:160997772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAAATACAATCTCACCAAAGAG[C/T]TAAGAAGCCCTCCAACACACGCCCA
Phenotype
MIM: 605721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
F11R PubMed Links

Gene Details

Gene
F11R
Gene Name
F11 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016946.4 2269 UTR 3 NP_058642.1

View Full Product Details