Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000120.3 | 1465 | Silent Mutation | CTG,TTG | L406L | NP_000111.1 |
NM_001136018.3 | 1465 | Silent Mutation | CTG,TTG | L406L | NP_001129490.1 |
NM_001291163.1 | 1465 | Silent Mutation | CTG,TTG | L406L | NP_001278092.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014698.2 | 1465 | Intron | NP_055513.2 | ||
XM_006711841.3 | 1465 | Intron | XP_006711904.1 | ||
XM_011544328.2 | 1465 | Intron | XP_011542630.1 | ||
XM_011544329.2 | 1465 | Intron | XP_011542631.1 | ||
XM_011544330.2 | 1465 | Intron | XP_011542632.1 | ||
XM_011544331.2 | 1465 | Intron | XP_011542633.1 | ||
XM_011544332.2 | 1465 | Intron | XP_011542634.1 | ||
XM_017002916.1 | 1465 | Intron | XP_016858405.1 |