Product Details
- SNP ID
-
rs1975213
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:26791402 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCATGAGTTTTTAATTTCCAATTAG[A/T]GAATTTGGCTTTTTTGATTCCTACT
- Phenotype
-
MIM: 117139
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CENPA
PubMed Links
Gene Details
- Gene
- CENPA
- Gene Name
- centromere protein A
View Full Product Details