Product Details
- SNP ID
-
rs1864536
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32357362 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGACGGCTGCATGCACTGCGACGC[C/T]GACGGGCTGCACAGCCTGTCCTACC
- Phenotype
-
MIM: 605638
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC6
PubMed Links
Gene Details
- Gene
- BIRC6
- Gene Name
- baculoviral IAP repeat containing 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016252.3 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
NP_057336.3 |
XM_005264449.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264506.2 |
XM_005264450.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264507.2 |
XM_005264451.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264508.2 |
XM_005264452.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264509.2 |
XM_005264453.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264510.2 |
XM_005264454.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264511.2 |
XM_005264455.4 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_005264512.2 |
XM_006712054.3 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_006712117.1 |
XM_006712055.3 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_006712118.1 |
XM_006712056.3 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_006712119.1 |
XM_011533003.2 |
456 |
Intron |
|
|
XP_011531305.1 |
XM_011533005.2 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_011531307.1 |
XM_017004556.1 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_016860045.1 |
XM_017004557.1 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_016860046.1 |
XM_017004558.1 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_016860047.1 |
XM_017004559.1 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_016860048.1 |
XM_017004560.1 |
456 |
Silent Mutation |
GCC,GCT |
A67A |
XP_016860049.1 |
View Full Product Details