Product Details

SNP ID
rs6701997
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:89057670 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAATATTAGTCCTTGAAATACAG[C/G]CATGTGAATTGACTTTTAAATTTTC
Phenotype
MIM: 600411
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GBP1 PubMed Links

Gene Details

Gene
GBP1
Gene Name
guanylate binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002053.2 Intron NP_002044.2

View Full Product Details