Product Details

SNP ID
rs3131828
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:92261583 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACTAGGAAGGGGTATGAGGAAC[A/C]TCTTGTGATGCTGGAAATGTTCTAT
Phenotype
MIM: 601749
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLMN PubMed Links

Gene Details

Gene
GLMN
Gene Name
glomulin, FKBP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319683.1 Intron NP_001306612.1
NM_053274.2 Intron NP_444504.1
XM_005270401.3 Intron XP_005270458.1
XM_006710309.2 Intron XP_006710372.1
XM_011540546.2 Intron XP_011538848.1
XM_017000137.1 Intron XP_016855626.1
XM_017000138.1 Intron XP_016855627.1
XM_017000139.1 Intron XP_016855628.1
XM_017000140.1 Intron XP_016855629.1
XM_017000141.1 Intron XP_016855630.1
XM_017000142.1 Intron XP_016855631.1
XM_017000143.1 Intron XP_016855632.1
XM_017000144.1 Intron XP_016855633.1

View Full Product Details