Product Details

SNP ID
rs13172376
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:6450398 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCAGGGTGAGTGCGTGATGGTGTTA[A/T]CTGCTCGAATTATATGCTGTCGACT
Phenotype
MIM: 615832
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2QL1 PubMed Links

Gene Details

Gene
UBE2QL1
Gene Name
ubiquitin conjugating enzyme E2 Q family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145161.2 Intron NP_001138633.1

View Full Product Details