Product Details

SNP ID
rs879597
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:31520730 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGTGCAGGGGTTGTGAAGACAGCA[A/G]TGCAGCAATGAGGGCCAAAGGATTG
Phenotype
MIM: 607106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HM13 PubMed Links

Gene Details

Gene
HM13
Gene Name
histocompatibility minor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030789.3 Intron NP_110416.1
NM_178580.2 Intron NP_848695.1
NM_178581.2 Intron NP_848696.1
NM_178582.2 Intron NP_848697.1

View Full Product Details