Product Details
- SNP ID
-
rs2000075
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:61282701 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCAAATTGCATTCTACAAGTCACA[C/T]TAAAATTAAAAAGTAATATTGATTT
- Phenotype
-
MIM: 605437
MIM: 611029
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRKCH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74412885] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRKCH
- Gene Name
- protein kinase C eta
- Gene
- TMEM30B
- Gene Name
- transmembrane protein 30B
View Full Product Details