Product Details

SNP ID
rs1952330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92524294 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCACTCATTCATTCACTGTCTCTT[C/T]GTTGAAACAGTCTTGGGGTCTCTTG
Phenotype
MIM: 610223
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIN3 PubMed Links

Gene Details

Gene
RIN3
Gene Name
Ras and Rab interactor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319987.1 Intron NP_001306916.1
NM_024832.4 Intron NP_079108.3
XM_011537164.2 Intron XP_011535466.1
XM_011537165.2 Intron XP_011535467.1
XM_017021651.1 Intron XP_016877140.1
XM_017021652.1 Intron XP_016877141.1
XM_017021653.1 Intron XP_016877142.1
XM_017021654.1 Intron XP_016877143.1

View Full Product Details