Product Details

SNP ID
rs11228732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56700679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCATCTCCTTTGCTGGCTGCCTG[G/T]GTCAGTTCTTCTTCTCTGCAGGGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR9G1 PubMed Links

Gene Details

Gene
OR9G1
Gene Name
olfactory receptor family 9 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005213.1 292 Missense Mutation GGT,TGT G98C NP_001005213.1

View Full Product Details