Product Details

SNP ID
rs2575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186149602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCAAGGATTATGCTCTGAGAGCC[A/G]GAGGCCGTTTTGTCTCTGTACAGAG
Phenotype
MIM: 601854
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKG PubMed Links

Gene Details

Gene
DGKG
Gene Name
diacylglycerol kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080744.1 3284 UTR 3 NP_001074213.1
NM_001080745.1 3284 UTR 3 NP_001074214.1
NM_001346.2 3284 UTR 3 NP_001337.2

View Full Product Details