Product Details

SNP ID
rs371296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153703238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTAGGCAAAGGCCAGGGCGTGG[C/T]TGCCCGCCGAGGGAAAGAGAAGCGC
Phenotype
MIM: 300398 MIM: 300036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAP31 PubMed Links

Gene Details

Gene
BCAP31
Gene Name
B-cell receptor-associated protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139441.1 Intron NP_001132913.1
NM_001139457.2 Intron NP_001132929.1
NM_001256447.1 Intron NP_001243376.1
NM_005745.7 Intron NP_005736.3
Gene
SLC6A8
Gene Name
solute carrier family 6 member 8
There are no transcripts associated with this gene.

View Full Product Details