Product Details

SNP ID
hCV11690968
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31270025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGCTCCAAGGACAGCTAGGACA[A/G]CCAGGACAGCCAGGCCAGCAACGAT
Phenotype
MIM: 142840
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HLA-C PubMed Links

Gene Details

Gene
HLA-C
Gene Name
major histocompatibility complex, class I, C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243042.1 1021 Missense Mutation GCT,GTT A319V NP_001229971.1
NM_002117.5 1021 Missense Mutation GCT,GTT A319V NP_002108.4

View Full Product Details