Product Details

SNP ID
rs4926222
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:14516238 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGGCTCAGCTCTTGCCCAGAGGCC[A/G]TCTGCCATAGATAAGACCCATCAGG
Phenotype
MIM: 604572
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB1 PubMed Links

Gene Details

Gene
DNAJB1
Gene Name
DnaJ heat shock protein family (Hsp40) member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300914.1 Intron NP_001287843.1
NM_001313964.1 Intron NP_001300893.1
NM_006145.2 Intron NP_006136.1
XM_006722733.1 Intron XP_006722796.1
XM_006722734.3 Intron XP_006722797.1
XM_011527956.2 Intron XP_011526258.1

View Full Product Details