Product Details
- SNP ID
-
rs1548476
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57547168 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCAGTGTGAATCCGCTGGTGCTGT[A/G]TGAGTTCTGAGCTGCATCTGAAGGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF549
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116624281] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF549
- Gene Name
- zinc finger protein 549
There are no transcripts associated with this gene.
- Gene
- ZNF550
- Gene Name
- zinc finger protein 550
View Full Product Details