Product Details

SNP ID
rs12610606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38275182 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACAAGCACGGTTCAAACTCCCCT[C/T]AGAAATTCTTTTGCACAGAAATTAA
Phenotype
MIM: 605124
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINT2 PubMed Links

Gene Details

Gene
SPINT2
Gene Name
serine peptidase inhibitor, Kunitz type 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166103.1 Intron NP_001159575.1
NM_021102.3 Intron NP_066925.1

View Full Product Details