Product Details

SNP ID
rs35835993
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35766210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTGGGGAATATGTCTTCTGGT[C/T]ACTGCTGTAACTCCAGTGTCCAGAA
Phenotype
MIM: 614902 MIM: 610695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP33 PubMed Links

Gene Details

Gene
ARHGAP33
Gene Name
Rho GTPase activating protein 33
There are no transcripts associated with this gene.

Gene
HSPB6
Gene Name
heat shock protein family B (small) member 6
There are no transcripts associated with this gene.

Gene
PROSER3
Gene Name
proline and serine rich 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039887.2 Intron NP_001034976.2
XM_011526528.1 Intron XP_011524830.1
XM_011526529.2 Intron XP_011524831.1
XM_011526530.1 Intron XP_011524832.1
XM_011526531.1 Intron XP_011524833.1
XM_011526532.1 Intron XP_011524834.1
XM_011526533.2 Intron XP_011524835.1
XM_011526534.1 Intron XP_011524836.1
XM_011526535.2 Intron XP_011524837.1
XM_011526536.2 Intron XP_011524838.1

View Full Product Details