Product Details
- SNP ID
-
rs31079
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55325229 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTCTGATCATCCTCCTCTTGTGT[C/T]CTGGGGGAAGCCAGAAGGAGCAAGG
- Phenotype
-
MIM: 606196
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IRX6
PubMed Links
Gene Details
- Gene
- IRX6
- Gene Name
- iroquois homeobox 6
View Full Product Details