Product Details
- SNP ID
-
rs2066841
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:56590999 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTGGAGATAGTGCCAGGTGGCTCA[G/T]TGCCACCTATGCCTGTGGTGAAGTG
- Phenotype
-
MIM: 139255
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MT3
PubMed Links
Gene Details
- Gene
- MT3
- Gene Name
- metallothionein 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005954.2 |
493 |
UTR 3 |
|
|
NP_005945.1 |
View Full Product Details