Product Details

SNP ID
rs13030612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:42172031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGAGCCATGAATATAAAGGCAAG[A/C]ACTTCTACTCTAGTAGGTGTTTAAA
Phenotype
MIM: 607442
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EML4 PubMed Links

Gene Details

Gene
EML4
Gene Name
echinoderm microtubule associated protein like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145076.2 Intron NP_001138548.1
NM_019063.4 Intron NP_061936.2
XM_005264267.1 Intron XP_005264324.1
XM_005264268.3 Intron XP_005264325.1
XM_006711991.1 Intron XP_006712054.1
XM_006711992.2 Intron XP_006712055.1
Gene
LOC102723824
Gene Name
uncharacterized LOC102723824
There are no transcripts associated with this gene.

View Full Product Details