Product Details
- SNP ID
-
rs8085532
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:31447808 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGCAGAGGGCTTTTCTTAGACAT[C/T]AACTGCAGACGGCTGGCAGGATAGA
- Phenotype
-
MIM: 169615
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DSG3
PubMed Links
Gene Details
- Gene
- DSG3
- Gene Name
- desmoglein 3
View Full Product Details