Product Details
- SNP ID
-
rs1980910
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:104643825 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATATATCCTAGGAAGAGGAGGATG[C/T]AATGTCTTTACATCTGTGGCTTCCC
- Phenotype
-
MIM: 606285
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SORCS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74938365] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SORCS3
- Gene Name
- sortilin related VPS10 domain containing receptor 3
View Full Product Details