Product Details
- SNP ID
-
rs1972895
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:67824156 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTGTGTTTTTTTTTTTTTTTTAA[T/G]TGTGAACTATGCATTATAGTATTTC
- Phenotype
-
MIM: 605369
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMPRSS11D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4572921,rs5859106] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMPRSS11D
- Gene Name
- transmembrane protease, serine 11D
View Full Product Details