Product Details

SNP ID
rs13250
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:44986581 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCCCAGGATGGCGGTGGATGACTA[A/G]ACTGCCGAAGACCGCGCCGCTGCGA
Phenotype
MIM: 600642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1G PubMed Links

Gene Details

Gene
MYO1G
Gene Name
myosin IG
There are no transcripts associated with this gene.

Gene
SNHG15
Gene Name
small nucleolar RNA host gene 15
There are no transcripts associated with this gene.

Gene
SNORA9
Gene Name
small nucleolar RNA, H/ACA box 9
There are no transcripts associated with this gene.

View Full Product Details