Product Details
- SNP ID
-
rs4075583
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:63048028 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTAGTGCTTGGGTTATAATCTTGC[A/G]GTTGGGATCAGGCCCTTTAGGGCAT
- Phenotype
-
MIM: 191010
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TPM1
PubMed Links
Gene Details
- Gene
- TPM1
- Gene Name
- tropomyosin 1 (alpha)
View Full Product Details