Product Details

SNP ID
rs8041223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59644327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGCAACACTTAAATGCCTAAAAT[C/G]TTCAACTCAGAAACCACCAAGCATG
Phenotype
MIM: 600519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTF2A2 PubMed Links

Gene Details

Gene
GTF2A2
Gene Name
general transcription factor IIA subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320929.1 Intron NP_001307858.1
NM_001320930.1 Intron NP_001307859.1
NM_004492.2 Intron NP_004483.1

View Full Product Details