Product Details
- SNP ID
-
rs10449043
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:70270566 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGATCTGGATCTTAGACTGCCTAT[A/G]GAATGATCCAGTAAGTCAGAAATCC
- Phenotype
-
MIM: 301770
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARR3
PubMed Links
Gene Details
- Gene
- ARR3
- Gene Name
- arrestin 3
View Full Product Details