Product Details

SNP ID
rs11921473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:131539911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTAGACACCTAGGGAGAAACTA[A/G]CTTCATTAATGAACATAATTGCAGC
Phenotype
MIM: 604208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPNE4 PubMed Links

Gene Details

Gene
CPNE4
Gene Name
copine 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289112.1 Intron NP_001276041.1
NM_130808.2 Intron NP_570720.1
NM_153429.1 Intron NP_702907.1
XM_011512406.2 Intron XP_011510708.1
XM_011512407.2 Intron XP_011510709.1
XM_011512408.2 Intron XP_011510710.1
XM_017005693.1 Intron XP_016861182.1
XM_017005694.1 Intron XP_016861183.1
XM_017005695.1 Intron XP_016861184.1
XM_017005696.1 Intron XP_016861185.1

View Full Product Details