Product Details
- SNP ID
-
rs6510810
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:500359 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATCCTGCCCACTGTGGTGCCTCAG[T/C]TTCCTCATCTGTCCAGTGGACTTCC
- Phenotype
-
MIM: 102670
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MADCAM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112902213] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MADCAM1
- Gene Name
- mucosal vascular addressin cell adhesion molecule 1
- Gene
- TPGS1
- Gene Name
- tubulin polyglutamylase complex subunit 1
There are no transcripts associated with this gene.
View Full Product Details