Product Details
- SNP ID
-
rs12625250
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62553197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGGCTGGTGGAGGCTGGGCGGC[C/T]GGCCACAAAGAGCCAGGTGTGGGAC
- Phenotype
-
MIM: 609326
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C20orf166-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117225122] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C20orf166-AS1
- Gene Name
- C20orf166 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MIR1-1
- Gene Name
- microRNA 1-1
There are no transcripts associated with this gene.
- Gene
- MIR1-1HG
- Gene Name
- MIR1-1 host gene
View Full Product Details