Product Details
- SNP ID
-
rs12837650
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:126821708 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGCACTAATGTGCCCTATTTGAAA[C/T]CTTGTACCCTACCACACTCTGTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR32
PubMed Links
Gene Details
- Gene
- PRR32
- Gene Name
- proline rich 32
View Full Product Details