Product Details

SNP ID
rs35887395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122740885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGTTTCCTCAACCTAAGGGAA[A/G]AGAAAAACCTTTTAAAATGGTTACA
Phenotype
MIM: 608263
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPBAP1 PubMed Links

Gene Details

Gene
HSPBAP1
Gene Name
HSPB1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320728.1 Intron NP_001307657.1
NM_024610.5 Intron NP_078886.2
XM_005247762.2 Intron XP_005247819.1
XM_011513130.2 Intron XP_011511432.1
XM_011513132.2 Intron XP_011511434.2
XM_017007179.1 Intron XP_016862668.1
XM_017007180.1 Intron XP_016862669.1

View Full Product Details