Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286708.1 | 2451 | UTR 3 | NP_001273637.1 | ||
NM_001286710.1 | 2451 | UTR 3 | NP_001273639.1 | ||
NM_001286711.1 | 2451 | UTR 3 | NP_001273640.1 | ||
NM_001286712.1 | 2451 | UTR 3 | NP_001273641.1 | ||
NM_001995.3 | 2451 | UTR 3 | NP_001986.2 | ||
XM_005262828.1 | 2451 | UTR 3 | XP_005262885.1 | ||
XM_005262829.1 | 2451 | UTR 3 | XP_005262886.1 | ||
XM_005262831.1 | 2451 | UTR 3 | XP_005262888.1 | ||
XM_011531742.1 | 2451 | UTR 3 | XP_011530044.1 | ||
XM_017007887.1 | 2451 | UTR 3 | XP_016863376.1 | ||
XM_017007888.1 | 2451 | UTR 3 | XP_016863377.1 | ||
XM_017007889.1 | 2451 | UTR 3 | XP_016863378.1 |