Product Details

SNP ID
rs2715755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108963371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGTTAGCTGAGTTCTCGTTCC[A/G]TAGAGTCTACTCCTACTGCTCAAAT
Phenotype
MIM: 605128 MIM: 603205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
There are no transcripts associated with this gene.

Gene
MORC1
Gene Name
MORC family CW-type zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014429.3 Intron NP_055244.3
XM_005247362.2 Intron XP_005247419.1
XM_011512691.1 Intron XP_011510993.1
XM_011512692.1 Intron XP_011510994.1
XM_011512693.1 Intron XP_011510995.1
XM_011512694.1 Intron XP_011510996.1
XM_011512696.2 Intron XP_011510998.1
XM_011512697.2 Intron XP_011510999.1
XM_017006169.1 Intron XP_016861658.1
XM_017006170.1 Intron XP_016861659.1

View Full Product Details