Product Details

SNP ID
rs12622321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:24030661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACACCATCCATATGGCACCATAA[C/T]ATTGTTTTGTTCTGAGAATTCTGCC
Phenotype
MIM: 600620 MIM: 616234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKBP1B PubMed Links

Gene Details

Gene
FKBP1B
Gene Name
FK506 binding protein 1B
There are no transcripts associated with this gene.

Gene
MFSD2B
Gene Name
major facilitator superfamily domain containing 2B
There are no transcripts associated with this gene.

Gene
WDCP
Gene Name
WD repeat and coiled coil containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142319.1 2569 UTR 3 NP_001135791.1
NM_025203.2 2569 UTR 3 NP_079479.1
XM_017005029.1 2569 Intron XP_016860518.1

View Full Product Details