Product Details

SNP ID
rs2955114
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:128158913 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAGAATTTGGCAAGTTTTTTATTT[A/C]GACTAAAACTGTAGAAGAACTTATT
Phenotype
MIM: 607695 MIM: 603449
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EEFSEC PubMed Links

Gene Details

Gene
EEFSEC
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021937.4 Intron NP_068756.2
XM_005247696.2 Intron XP_005247753.1
XM_011513066.2 Intron XP_011511368.1
XM_017006975.1 Intron XP_016862464.1
XM_017006976.1 Intron XP_016862465.1
XM_017006977.1 Intron XP_016862466.1
XM_017006978.1 Intron XP_016862467.1
XM_017006979.1 Intron XP_016862468.1
XM_017006980.1 Intron XP_016862469.1
XM_017006981.1 Intron XP_016862470.1
Gene
RUVBL1
Gene Name
RuvB like AAA ATPase 1
There are no transcripts associated with this gene.

View Full Product Details