Product Details

SNP ID
rs10772998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123236630 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAAGAAGAAACTGCTTTGGTTTG[G/T]ATATGTTTAAGTCCTAAACAAACAC
Phenotype
MIM: 613541 MIM: 605501
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf65 PubMed Links

Gene Details

Gene
C12orf65
Gene Name
chromosome 12 open reading frame 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143905.2 Intron NP_001137377.1
NM_001194995.1 Intron NP_001181924.1
NM_152269.4 Intron NP_689482.1
XM_005253630.4 Intron XP_005253687.1
XM_011538980.2 Intron XP_011537282.1
Gene
MPHOSPH9
Gene Name
M-phase phosphoprotein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022782.3 Intron NP_073619.3
XM_011537740.2 Intron XP_011536042.1
XM_011537741.2 Intron XP_011536043.1
XM_017018673.1 Intron XP_016874162.1
XM_017018674.1 Intron XP_016874163.1
XM_017018675.1 Intron XP_016874164.1
XM_017018676.1 Intron XP_016874165.1
XM_017018677.1 Intron XP_016874166.1
XM_017018678.1 Intron XP_016874167.1
XM_017018679.1 Intron XP_016874168.1
XM_017018680.1 Intron XP_016874169.1
XM_017018681.1 Intron XP_016874170.1

View Full Product Details