Product Details
- SNP ID
-
rs2280390
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:186576031 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCATTCCCTAGTTCAGAAACTTT[T/G]GATGAACAGTACAATTTAGCTGATC
- Phenotype
-
MIM: 611341
- Polymorphism
- T/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAJB11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56227532] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAJB11
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016306.5 |
|
Intron |
|
|
NP_057390.1 |
- Gene
- TBCCD1
- Gene Name
- TBCC domain containing 1
There are no transcripts associated with this gene.
View Full Product Details